missing translation for 'onlineSavingsMsg'
Learn More

[3,4]-Dibenzo-21-crown-7, 98%, Thermo Scientific Chemicals

Catalog Number p-101105
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14098-41-0
C22H28O7
404.45
MFCD00068646
JKCQOMAQPUYHPL-UHFFFAOYSA-N
dibenzo-21-crown-7, dibenzo-21-crown 7-ether, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine, dibenzo-21-crown7-ether, dibenzo-21-crown, 2,3,14,15-dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosa-2,14-diene, 2,5,8,15,18,21,24-heptaoxatricyclo 23.4.0.0?, 1 ? nonacosa-1 29 ,9,11,13,25,27-hexaene, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine #, dibenzo b,k 1,4,7,10,13,16,19 heptaoxacycloheneicosin,6,7,9,10,12,13,20,21,23,24-decahydro
585932
C1COCCOC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
This item is not returnable. View return policy
14098-41-0
C22H28O7
404.45
MFCD00068646
JKCQOMAQPUYHPL-UHFFFAOYSA-N
dibenzo-21-crown-7, dibenzo-21-crown 7-ether, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine, dibenzo-21-crown7-ether, dibenzo-21-crown, 2,3,14,15-dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosa-2,14-diene, 2,5,8,15,18,21,24-heptaoxatricyclo 23.4.0.0?, 1 ? nonacosa-1 29 ,9,11,13,25,27-hexaene, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine #, dibenzo b,k 1,4,7,10,13,16,19 heptaoxacycloheneicosin,6,7,9,10,12,13,20,21,23,24-decahydro
585932
C1COCCOC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 104.0°C to 108.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 404.45
Percent Purity 98%
Physical Form Fluffy Amorphous Powder
Chemical Name or Material [3,4]-Dibenzo-21-crown-7, 98%
Show More Show Less
compliance-icons
Product Identifier
  • ¢3,4!-Dibenzo-21-crown-7
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'