missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminoanisole dihydrochloride, 98%, Acros Organics

Brand:  Acros Organics 396990250

Code : 29

Additional Details : CAS Number : 59548-39-9 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 25g

 View more versions of this product

Product Code. 10614755

  • £219.00 / 25g


Estimated Shipment
18-05-2018
Add to basket


Specifications

Chemical Identifiers

3, 4-Diaminoanisole dihydrochloride
98%
C7H10N22ClH
211.09

Specifications

3, 4-Diaminoanisole dihydrochloride
25g
Glass bottle
98%
C7H10N22ClH
211.09
206.0° to 209.0°C
59548-39-9
Solubility in water: soluble.
98%
CH3OC6H3(NH2)2·2HCl
Videos
SDS