missing translation for 'onlineSavingsMsg'
Learn More

3,4-Di-O-acetyl-6-O-tert-butyldimethylsilyl-D-glucal, 97%, Thermo Scientific™

Catalog Number 15436086
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
117136-33-1
C16H28O6Si
MFCD01321277
3,4-di-o-acetyl-6-o-tert-butyldimethylsilyl-d-glucal, 3beta,4alpha-diacetoxy-2alpha-tert-butyldimethylsiloxymethyl-3,4-dihydro-2h-pyran, 2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 2r,3s,4r-4-acetyloxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3-yl acetate
This item is not returnable. View return policy
117136-33-1
C16H28O6Si
MFCD01321277
3,4-di-o-acetyl-6-o-tert-butyldimethylsilyl-d-glucal, 3beta,4alpha-diacetoxy-2alpha-tert-butyldimethylsiloxymethyl-3,4-dihydro-2h-pyran, 2r,3s,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 2r,3s,4r-4-acetyloxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3-yl acetate

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.044
Flash Point >113°C (235°F)
Refractive Index 1.456
Quantity 250 mg
Optical Rotation −7° (c=1.4 in Chloroform)
Formula Weight 344.48
Percent Purity 97%
Chemical Name or Material 3,4-Di-O-acetyl-6-O-tert-butyldimethylsilyl-D-glucal
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'