Learn More
Thermo Scientific Maybridge 3-(4-Chlorophenyl)propan-1-ol, 1g, 95%
Brand: Thermo Scientific Maybridge KM10071DA
3-(4-Chlorophenyl)propan-1-ol, 1g, 95%
Code : 31
Additional Details : CAS Number : 6282-88-8 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge KM10071DA
3-(4-Chlorophenyl)propan-1-ol, 1g, 95%
Code : 31
Additional Details : CAS Number : 6282-88-8 Weight : 0.05100kg