missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Chlorophenyl)glutaric acid, 98%

Brand:  Alfa Aesar™ L18693.14

Code : NEW

Additional Details : CAS Number : 35271-74-0 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11369465

  • £72.80 / 25g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Description

Description

Applications
It is employed in an efficient synthesis of (R)- and (S)-baclofen via desymmetrization.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

3-(4-Chlorophenyl)glutaric acid
C11H11ClO4
MFCD00190249
35271-74-0
242.66
1976828

Specifications

3-(4-Chlorophenyl)glutaric acid
25g
98%
242.66
1976828
164° to 168°C
35271-74-0
C11H11ClO4
MFCD00190249
Videos
SDS