missing translation for 'onlineSavingsMsg'
Learn More

3-(4-chlorophenyl)-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Catalog Number p-2699084
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
78583-81-0
C9H8ClN3
193.63
MFCD00053046
XQPBZIITFQHIDI-UHFFFAOYSA-N
3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
2735305
5-(4-chlorophenyl)-1H-pyrazol-3-amine
NC1=NNC(=C1)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
78583-81-0
C9H8ClN3
193.63
MFCD00053046
XQPBZIITFQHIDI-UHFFFAOYSA-N
3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
2735305
5-(4-chlorophenyl)-1H-pyrazol-3-amine
NC1=NNC(=C1)C1=CC=C(Cl)C=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.64
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenyl)-1H-pyrazol-5-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'