missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Synonym | 3-4-bromophenyl-5-methyl-1,2,4-oxadiazole, 4-5-methyl-1,2,4-oxadiazol-3-yl bromobenzene, acmc-1ce2y, 3-4-bromophenyl-5-methyl-1,2,4oxadiazole, 1,2,4-oxadiazole, 3-4-bromophenyl-5-methyl, 1,2,4-oxadiazole,3-4-bromophenyl-5-methyl, 3-4-bromo-phenyl-5-methyl-1,2,4 oxadiazole, 3-4-bromophenyl-5-methyl-1,2,4 oxadiazole, 1-bromo-4-5-methyl-1,2,4-oxadiazol-3-yl benzene |
Formula Weight | 239.07 |
Percent Purity | 97% |
Chemical Name or Material | 3-(4-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'