missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Bromophenyl)-2-thioxo-4-thiazolidinone, 95%

Brand:  Alfa Aesar™ H63281.06

Code : NEW

Additional Details : CAS Number : 10574-70-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15479898

  • £498.20 / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Bromophenyl)-2-thioxo-4-thiazolidinone
288.18
3-(4-Bromophenyl)rhodanine
10574-70-6
MFCD00174856

Specifications

3-(4-Bromophenyl)-2-thioxo-4-thiazolidinone
5g
95%
MFCD00174856
10574-70-6
Solid
288.18
3-(4-Bromophenyl)rhodanine
Videos
SDS