missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Bromophenyl)-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63309.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459938

  • £62.96 / 1g


Estimated Shipment date
from Supplier 14-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Bromophenyl)-1-phenyl-2-thiohydantoin
347.23
MFCD27976891
3-(4-Bromophenyl)-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-(4-Bromophenyl)-1-phenyl-2-thiohydantoin
Solid
347.23
3-(4-Bromophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD27976891
Videos
SDS