missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Bromophenyl)-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63044.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419498

  • £373.12 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Bromophenyl)-1-methyl-2-thiohydantoin
285.16
MFCD27665227
3-(4-Bromophenyl)-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-(4-Bromophenyl)-1-methyl-2-thiohydantoin
Solid
285.16
3-(4-Bromophenyl)-1-methyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD27665227
Videos
SDS