missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[4-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 253.1 |
Percent Purity | 97% |
Chemical Name or Material | 3-[4-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'