missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromobenzoyl)propionic acid, 97%, ACROS Organics™

Brand:  Acros Organics 344340250

£58.16 valid until 30/06/2017
Log in to get the promotional price



UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 6340-79-0 Weight : 0.08000kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10061861

  • £145.40 / 25g


EU Stock 1
Add to basket


Specifications Description & Specifications

Chemical Identifiers

3-(4-Bromobenzoyl)propionic acid
96.0
≥96.0%
C10H9BrO3
257.08
6340-79-0
100.0
97%
BrC6H4CO(CH)COOH
MFCD00016563

Specifications

3-(4-Bromobenzoyl)propionic acid
Authentic
100.0
White to light beige
Shiny Crystalline Flakes or Powder
97%
BrC6H4CO(CH)COOH
MFCD00016563
6340-79-0
96.0
Glass bottle
25g
≥96.0%
C10H9BrO3
257.08

Safety and Handling

Product Identifier
  • 3-(4-Bromobenzoyl)propionic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents

Special Offers

Special Offers