missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(4-Bromo-3-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Brand: Alfa Aesar™ H56261.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
3-(4-Bromo-3-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid | |
370.96 | |
MFCD27665220 |
Specifications
3-(4-Bromo-3-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid | |
Solid | |
370.96 |
1g | |
97% | |
MFCD27665220 |