missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Bromo-2-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56094.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15472476

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Bromo-2-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid
370.96
MFCD27665218

Specifications

3-(4-Bromo-2-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid
Solid
370.96
1g
97%
MFCD27665218
Videos
SDS