missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034269
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
883738-19-0
C22H35BN2O4
402.342
MFCD16294542
KBHWOVSSXKGKLY-UHFFFAOYSA-N
tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, 3-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, amtb241, 3-4-boc-piperazine methyl phenylboronic acid pinacol ester, 3-4-boc-piperazine methyl phenylboronic-acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl-1-piperazinecarboxylic acid tert-butyl ester
59298946
tert-butyl 4-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CN3CCN(CC3)C(=O)OC(C)(C)C
This item is not returnable. View return policy
883738-19-0
C22H35BN2O4
402.342
MFCD16294542
KBHWOVSSXKGKLY-UHFFFAOYSA-N
tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperazine-1-carboxylate, 3-4-boc-1-piperazinylmethyl benzeneboronic acid pinacol ester, tert-butyl 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperazine-1-carboxylate, amtb241, 3-4-boc-piperazine methyl phenylboronic acid pinacol ester, 3-4-boc-piperazine methyl phenylboronic-acid pinacol ester, 4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl-1-piperazinecarboxylic acid tert-butyl ester
59298946
tert-butyl 4-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperazine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CN3CCN(CC3)C(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Product of Advanced Molecular Technologies Pty Ltd.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 402.34
Percent Purity 95%
Chemical Name or Material 3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'