missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzaldehyde, 99%, ACROS Organics™

Brand:  Acros Organics 139911000

Code : 29

Additional Details : CAS Number : 86-81-7 Weight : 0.15000kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10562151

  • £33.60 / 100g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

3, 4, 5-Trimethoxybenzaldehyde
98.5
99%
C10H12O4
196.2
86-81-7
100.0
99%
(CH3O)3C6H2CHO
MFCD00003364

Specifications

3, 4, 5-Trimethoxybenzaldehyde
White to creamy
100g
98.5
Authentic
Powder or Flakes
99%
(CH3O)3C6H2CHO
MFCD00003364
163.0° to 165.0°C (10.0 mmHg)
73.0° to 76.0°C
86-81-7
100.0
Glass bottle
99%
C10H12O4
196.2
08, 391
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4,5-Trimethoxybenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS