missing translation for 'onlineSavingsMsg'
Learn More

3',4',5'-Trimethoxyacetophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-7058313
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1136-86-3
C11H14O4
210.23
MFCD00008738
VUGQIIQFXCXZJU-UHFFFAOYSA-N
3',4',5'-trimethoxyacetophenone, 1-3,4,5-trimethoxyphenyl ethanone, 3,4,5-trimethoxyacetophenone, ethanone, 1-3,4,5-trimethoxyphenyl, 1-3,4,5-trimethoxyphenyl ethan-1-one, 3,4,5-trimethoxy acetophenone, pubchem22820, acmc-1c5os, 3,5-trimethoxy acetophenone, 1-acetyl-3,4,5-trimethoxybenzene
14345
CHEBI:86547
1-(3,4,5-trimethoxyphenyl)ethanone
COC1=CC(=CC(OC)=C1OC)C(C)=O
This item is not returnable. View return policy
1136-86-3
C11H14O4
210.23
MFCD00008738
VUGQIIQFXCXZJU-UHFFFAOYSA-N
3',4',5'-trimethoxyacetophenone, 1-3,4,5-trimethoxyphenyl ethanone, 3,4,5-trimethoxyacetophenone, ethanone, 1-3,4,5-trimethoxyphenyl, 1-3,4,5-trimethoxyphenyl ethan-1-one, 3,4,5-trimethoxy acetophenone, pubchem22820, acmc-1c5os, 3,5-trimethoxy acetophenone, 1-acetyl-3,4,5-trimethoxybenzene
14345
CHEBI:86547
1-(3,4,5-trimethoxyphenyl)ethanone
COC1=CC(=CC(OC)=C1OC)C(C)=O

3',4',5'-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′,5′-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Slightly soluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store away from strong oxidizing agents and stong bases. Avoid excessive heat and light exposure.
TRUSTED_SUSTAINABILITY
Melting Point 79°C to 83°C
Boiling Point 173°C to 174°C (10 mmHg)
Quantity 5 g
Beilstein 1463133
Solubility Information Slightly soluble in water.
Formula Weight 210.23
Percent Purity 99%
Chemical Name or Material 3',4',5'-Trimethoxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'