missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',4',5'-Trimethoxyacetophenone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3',4',5'-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′,4′,5′-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Slightly soluble in water.
Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store away from strong oxidizing agents and stong bases. Avoid excessive heat and light exposure.
3′,4′,5′-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Slightly soluble in water.
Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store away from strong oxidizing agents and stong bases. Avoid excessive heat and light exposure.
Specifications
Melting Point | 79°C to 83°C |
Boiling Point | 173°C to 174°C (10 mmHg) |
Quantity | 5 g |
Beilstein | 1463133 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 210.23 |
Percent Purity | 99% |
Chemical Name or Material | 3',4',5'-Trimethoxyacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'