missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorophenylboronic acid, 95+%, Thermo Scientific™

Catalog Number p-3779770
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
143418-49-9
C6H4BF3O2
175.901
MFCD02093069
UHDDEIOYXFXNNJ-UHFFFAOYSA-N
3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj
2734671
(3,4,5-trifluorophenyl)boronic acid
B(C1=CC(=C(C(=C1)F)F)F)(O)O
This item is not returnable. View return policy
143418-49-9
C6H4BF3O2
175.901
MFCD02093069
UHDDEIOYXFXNNJ-UHFFFAOYSA-N
3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj
2734671
(3,4,5-trifluorophenyl)boronic acid
B(C1=CC(=C(C(=C1)F)F)F)(O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 175.9
Percent Purity ≥95.0%
Physical Form Powder
Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4,5-Trifluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'