missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorophenol, 98%, ACROS Organics™

Brand:  Acros Organics 442580010

Code : 29

Additional Details : CAS Number : 99627-05-1 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 1a Pack group : II


Quantity 1g

 View more versions of this product

Product Code. 11994681

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3, 4, 5-Trifluorophenol
97.5
98%
C6H3F3O
99627-05-1
100.0
98%
148.09

Specifications

3, 4, 5-Trifluorophenol
50.0° to 51.0°C
99627-05-1
97.5
Crystals or Low Melting Solid
98%
148.09
Colorless to pale yellow
1g
Authentic
100.0
98%
C6H3F3O
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4,5-Trifluorophenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS