missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trifluorobenzyl alcohol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3,4,5-Trifluorobenzyl alcohol is used as chemical reagent.
Solubility
Soluble in water 100 g/L.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
3,4,5-Trifluorobenzyl alcohol is used as chemical reagent.
Solubility
Soluble in water 100 g/L.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Density | 1.385 |
Boiling Point | 203°C to 204°C |
Flash Point | 98°C (208°F) |
Refractive Index | 1.4725 |
Quantity | 1 g |
Beilstein | 8681069 |
Synonym | 3,4,5-trifluorobenzyl alcohol, 3,4,5-trifluorophenyl methanol, 3,4,5-trifluorobenzenemethanol, benzenemethanol, 3,4,5-trifluoro, 3,4,5-trifluorophenyl methan-1-ol, 3,4,5-trifluoro benzenemethanol, acmc-20angf, 3,4,5-trifluorobenzylalcohol, 3,4,5-trifluoro-benzenemethanol, benzenemethanol,3,4,5-trifluoro |
Solubility Information | Soluble in water 100g/L. |
Formula Weight | 162.11 |
Percent Purity | 97% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'