missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3',4',5'-Trifluoroacetophenone, 97%

Brand:  Alfa Aesar™ L19187.06

Code : NEW

Additional Details : CAS Number : 220141-73-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11304066

  • £137.60 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

3',4',5'-Trifluoroacetophenone
C8H5F3O
MFCD00083533
220141-73-1
174.12

Specifications

3',4',5'-Trifluoroacetophenone
5g
97%
174.12
>100°C (212°F)
220141-73-1
C8H5F3O
MFCD00083533
Videos
SDS