missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5,6-Tetrahydrophthalic anhydride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 70.0°C to 74.0°C |
Color | White to Cream |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass Bottle |
Quantity | 100 g |
Beilstein | 17,461 |
Solubility Information | Solubility in water: reacts. |
IUPAC Name | 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione |
Formula Weight | 152.15 |
Show More |
Safety and Handling
Product Identifier
- 3,4,5,6-Tetrahydrophthalic anhydride
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'