missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, ACROS Organics™

Brand:  Acros Organics 366410010

Code : 29

Additional Details : CAS Number : 214360-76-6 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10571102

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
214360-76-6
100.0
97%
220.08
97%
96.0
97%
C12H17BO3
MFCD02093755

Specifications

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
90.0° to 96.0°C
97%
96.0
Authentic
Crystalline Powder
97%
220.08
Solubility in water: insoluble in water.
Light brown
1g
214360-76-6
100.0
Glass bottle
97%
C12H17BO3
MFCD02093755
Videos
SDS