missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, ACROS Organics™

Brand:  Acros Organics 367570010

Code : 29

Additional Details : CAS Number : 269409-73-6 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10591102

  • £19.50 / 1g


Estimated Shipment
14-06-2018
Add to basket


Specifications

Chemical Identifiers

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
269409-73-6
100.0
97%
248.09
3-Carboxyphenylboronic acid pinacol ester
97%
96.0
97%
C13H17BO4
MFCD03411930

Specifications

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
206.0° to 211.0°C
97%
96.0
Authentic
Crystalline Powder
97%
248.09
Solubility in water: insoluble in water.
Almost white to light beige
1g
269409-73-6
100.0
Glass bottle
97%
C13H17BO4
MFCD03411930
3-Carboxyphenylboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS