Alfa Aesar™ 3-[3-(Trifluoromethyl)phenyl]propanol, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64553.MD

250MG 3-[3-(Trifluoromethyl)phenyl]propanol, 98%250mg

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 78573-45-2 Weight: 0.00025kg


Quantity 250mg

Product Code. 15461829

Quantity Price
1 £ 13.99 / 250mg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propanol
CAS 78573-45-2
Formula Weight 204.19
MDL Number MFCD08706408

Specification

Formula Weight 204.19
Boiling Point 68° to 71°C (0.4mmHg)
Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propanol
Physical Form Liquid
Assay 98%
MDL Number MFCD08706408
CAS 78573-45-2
Density 1.10g/mL
Color Colorless
Quantity 250mg