missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals
360.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.
Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.
Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 248°C to 252°C (decomposition) |
Color | White |
Beilstein | 4351583 |
Solubility Information | Soluble in clear solution (0.3 gram in 2 ml DMF). |
Formula Weight | 166.18 |
Percent Purity | 98% |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | 3-(3-Pyridyl)-L-alanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'