missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Hydroxyphenyl)-DL-alanine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 1M HCl.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 1M HCl.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 280°C to 285°C (decomposition) |
Color | White to Yellow |
Beilstein | 2416853 |
Merck Index | 14,9840 |
Solubility Information | Soluble in 1M HCl. |
Formula Weight | 181.19 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | 3-(3-Hydroxyphenyl)-DL-alanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'