missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Hydroxyphenyl)-DL-alanine, 98%, Thermo Scientific Chemicals

Catalog Number 15459698
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
775-06-4
C9H11NO3
181.19
MFCD00002597
JZKXXXDKRQWDET-UHFFFAOYNA-N
dl-m-tyrosine, 2-amino-3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl-dl-alanine, m-tyrosine, dl, d,l-metatyrosine, 3-hydroxy-dl-phenylalanine, dl-meta-tyrosine, dl-phenylalanine, 3-hydroxy, 3-m-hydroxyphenyl-dl-alanine, dl-3-tyrosine
13052
2-amino-3-(3-hydroxyphenyl)propanoic acid
NC(CC1=CC=CC(O)=C1)C(O)=O
This item is not returnable. View return policy
775-06-4
C9H11NO3
181.19
MFCD00002597
JZKXXXDKRQWDET-UHFFFAOYNA-N
dl-m-tyrosine, 2-amino-3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl-dl-alanine, m-tyrosine, dl, d,l-metatyrosine, 3-hydroxy-dl-phenylalanine, dl-meta-tyrosine, dl-phenylalanine, 3-hydroxy, 3-m-hydroxyphenyl-dl-alanine, dl-3-tyrosine
13052
2-amino-3-(3-hydroxyphenyl)propanoic acid
NC(CC1=CC=CC(O)=C1)C(O)=O

3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1M HCl.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 280°C to 285°C (decomposition)
Color White to Yellow
Beilstein 2416853
Merck Index 14,9840
Solubility Information Soluble in 1M HCl.
Formula Weight 181.19
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material 3-(3-Hydroxyphenyl)-DL-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'