missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Fluorophenyl)-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63352.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410009

  • £79.71 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(3-Fluorophenyl)-2-thiohydantoin
210.23
MFCD27665231
3-(3-Fluorophenyl)-2-thioxo-4-imidazolidinone

Specifications

3-(3-Fluorophenyl)-2-thiohydantoin
Solid
210.23
3-(3-Fluorophenyl)-2-thioxo-4-imidazolidinone
1g
95%
MFCD27665231
Videos
SDS