Alfa Aesar™ 3-(3-Fluorophenyl)-2-thiohydantoin, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H63352.06

UNSPSC: 12352115

Code: NEW

Additional Details:
Additional Details: Weight: 0.00500kg


Quantity 5g

Product Code. 15420009

Quantity Price
1 £ 398.56 / 5g
Estimated Shipment date
from Supplier 11-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-(3-Fluorophenyl)-2-thiohydantoin
Formula Weight 210.23
MDL Number MFCD27665231
Synonym 3-(3-Fluorophenyl)-2-thioxo-4-imidazolidinone

Specification

Formula Weight 210.23
MDL Number MFCD27665231
Chemical Name or Material 3-(3-Fluorophenyl)-2-thiohydantoin
Assay 95%
Synonym 3-(3-Fluorophenyl)-2-thioxo-4-imidazolidinone
Quantity 5g
Physical Form Solid
Documentation

Technical documents