missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Fluorophenyl)-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64633.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15431919

  • £271.36 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(3-Fluorophenyl)-1-phenyl-2-thiohydantoin
286.32
MFCD27665296
3-(3-Fluorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-(3-Fluorophenyl)-1-phenyl-2-thiohydantoin
Solid
286.32
3-(3-Fluorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD27665296
Videos
SDS