missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile, 97%

Brand:  Alfa Aesar™ H57516.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15423978

  • £107.70 / 1g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile
199.21
MFCD22682912
UN3439

Specifications

4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile
64° to 66°C
Solid
199.21
UN3439
White to off-white
1g
97%
MFCD22682912
Videos
SDS