missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-124221
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1119-62-6
C6H10O4S2
210.26
MFCD00002780
YCLSOMLVSHPPFV-UHFFFAOYSA-N
3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid
95116
3-(2-carboxyethyldisulfanyl)propanoic acid
C(CSSCCC(=O)O)C(=O)O
This item is not returnable. View return policy
1119-62-6
C6H10O4S2
210.26
MFCD00002780
YCLSOMLVSHPPFV-UHFFFAOYSA-N
3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid
95116
3-(2-carboxyethyldisulfanyl)propanoic acid
C(CSSCCC(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98
Melting Point 155°C to 158°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula S2(CH2CH2CO2H)2
Quantity 50 g
Beilstein 03, IV, 736
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,benzene
Formula Weight 210.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 3'-Dithiodipropionic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3'-Dithiodipropionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'