missing translation for 'onlineSavingsMsg'
Learn More
Learn More
{3-[3-(Dimethylamino)propoxy]phenyl}methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 912569-56-3 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C12H19NO2 |
MDL Number | MFCD09064989 |
Quantity | 5 g |
Synonym | 3-3-dimethylamino propoxy phenyl methanol, benzenemethanol, 3-3-dimethylamino propoxy, 3-3-dimethylamino-propoxy-phenyl-methanol, 3-3-dimethylamino propoxy phenyl methan-1-ol |
InChI Key | BABQTNDHVRVEJD-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'