missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-2-butanol, 98+%, Thermo Scientific Chemicals

Catalog Number 11300338
Click to view available options
Quantity:
10 g
5 x 10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
464-07-3
C6H14O
102.177
MFCD00004522
DFOXKPDFWGNLJU-UHFFFAOYSA-N
3,3-dimethyl-2-butanol, pinacolyl alcohol, 2-butanol, 3,3-dimethyl, 2,2-dimethyl-3-butanol, tert-butyl methyl carbinol, 3,3-dimethylbutane-2-ol, 2-butanol, 3,3-dimethyl-, s, s-3,3-dimethyl-2-butanol, pinacolone alcohol, 3,3-dimethyl 2-butanol
10045
3,3-dimethylbutan-2-ol
CC(C(C)(C)C)O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
464-07-3
C6H14O
102.177
MFCD00004522
DFOXKPDFWGNLJU-UHFFFAOYSA-N
3,3-dimethyl-2-butanol, pinacolyl alcohol, 2-butanol, 3,3-dimethyl, 2,2-dimethyl-3-butanol, tert-butyl methyl carbinol, 3,3-dimethylbutane-2-ol, 2-butanol, 3,3-dimethyl-, s, s-3,3-dimethyl-2-butanol, pinacolone alcohol, 3,3-dimethyl 2-butanol
10045
3,3-dimethylbutan-2-ol
CC(C(C)(C)C)O

3,3-Dimethyl-2-butanol was used in conversion of ribose- and glucose-binding proteins into receptors for pinacolyl methyl phosphonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 5.3°C
Density 0.812g/mL
Boiling Point 119°C to 121°C
Flash Point 28°C (84°F)
Refractive Index 1.4150
Quantity 10 g
UN Number UN1987
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-2-butanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'