missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethoxypropanenitrile, 90%, Thermo Scientific Chemicals
104.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.0260g/mL |
Boiling Point | 195°C |
Flash Point | 83°C |
Assay Percent Range | 88% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3O)2CHCH2CN |
Refractive Index | 1.4130 to 1.415 |
Quantity | 25 mL |
Specific Gravity | 1.026 |
Synonym | 3,3-dimethoxypropionitrile, propanenitrile, 3,3-dimethoxy, 3,3-dimethoxypropiononitrile, 3,3-dimethoxy propionitrile, 3,3-dimethoxy-propionitrile, 3,3-dimethoxy-propanenitrile, 3,3-dimethoxypropanitrile, acmc-209lz7, #, propanenitrile,3,3-dimethoxy |
Show More |
Safety and Handling
Product Identifier
- 3,3-Dimethoxy-propanenitrile
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'