missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3,3'-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 81°C to 83°C |
Quantity | 1 g |
Beilstein | 2382124 |
Solubility Information | Insoluble in water. |
Formula Weight | 270.29 |
Percent Purity | ≥99% |
Chemical Name or Material | 3,3'-Dimethoxybenzil |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'