Learn More
Alfa Aesar™ 3,3-Dimethoxy-2-butanone, 98%
Brand: Alfa Aesar™ H64365.06
Code : NEW
Additional Details : CAS Number : 21983-72-2 Weight : 0.00500kg Transport : UN number : 1224 Chem class : 1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
3,3-Dimethoxy-2-butanone | |
132.16 | |
UN1224 |
21983-72-2 | |
MFCD00075266 | |
1752768 |
Specifications
3,3-Dimethoxy-2-butanone | |
1.4070 | |
0.987g/mL | |
145° to 146°C | |
132.16 | |
UN1224 | |
5g |
21983-72-2 | |
Liquid | |
98% | |
Colorless to pale yellow | |
MFCD00075266 | |
1752768 |