missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diethoxy-1-propanol, 98%, Thermo Scientific Chemicals

Catalog Number p-3770984
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16777-87-0
C7H16O3
148.20
MFCD00074850
ASERXEZXVIJBRO-UHFFFAOYSA-N
140135
3,3-diethoxypropan-1-ol
CCOC(CCO)OCC
This item is not returnable. View return policy
16777-87-0
C7H16O3
148.20
MFCD00074850
ASERXEZXVIJBRO-UHFFFAOYSA-N
140135
3,3-diethoxypropan-1-ol
CCOC(CCO)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9410g/mL
Boiling Point 90°C to 93°C (14.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2O)2CHCH2CH2OH
Quantity 1 g
Specific Gravity 0.941
Synonym 3,3-diethoxy-1-propanol, 3,3-diethoxypropanol, 1-propanol, 3,3-diethoxy, 3-hydroxypropionaldehyde diethyl acetal, 3, 3-diethoxypropanol, acmc-1c5mk, 1-propanol,3,3-diethoxy
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 3-Diethoxy-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 3,3-Diethoxy-1-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'