missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,3'-Di-n-hexyloxacarbocyanine iodide, 98%

Brand:  Alfa Aesar™ H55574.03

Code : NEW

Additional Details : CAS Number : 53213-82-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15431878

  • £100.60 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3,3'-Di-n-hexyloxacarbocyanine iodide
572.53
9312562
53213-82-4
MFCD00011954

Specifications

3,3'-Di-n-hexyloxacarbocyanine iodide
219° to 221°C
Red
Powder
572.53
9312562
53213-82-4
1g
Light Sensitive
98%
MFCD00011954
Videos
SDS