missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-n-hexyloxacarbocyanine iodide, 98%, Thermo Scientific Chemicals

Catalog Number 15441878
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
53213-82-4
C29H37IN2O2
572.53
MFCD00011954
XVLXYDXJEKLXHN-UHFFFAOYSA-M
3,3'-dihexyloxacarbocyanine iodide
129893825
[I-].CCCCCCN1\C(OC2=CC=CC=C12)=C\C=CC1=[N+](CCCCCC)C2=CC=CC=C2O1
This item is not returnable. View return policy
53213-82-4
C29H37IN2O2
572.53
MFCD00011954
XVLXYDXJEKLXHN-UHFFFAOYSA-M
3,3'-dihexyloxacarbocyanine iodide
129893825
[I-].CCCCCCN1\C(OC2=CC=CC=C12)=C\C=CC1=[N+](CCCCCC)C2=CC=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 219°C to 221°C
Color Red
Quantity 5 g
Beilstein 9312562
Sensitivity Light sensitive
IUPAC Name (2Z)-3-hexyl-2-[(E)-3-(3-hexyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
Formula Weight 572.53
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3,3'-Di-n-hexyloxacarbocyanine iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'