missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56705.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15442846

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(3-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
293.48
MFCD22682884

Specifications

3-(3-Chlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
Solid
293.48
1g
97%
MFCD22682884
Videos
SDS