missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Chloro-4-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56169.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402526

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(3-Chloro-4-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid
307.52
MFCD22682842

Specifications

3-(3-Chloro-4-methylphenylcarbamoyl)-2-fluorobenzeneboronic acid
97%
1g
MFCD22682842
Solid
Pale yellow
307.52
Videos
SDS