missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Bromophenyl)-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63261.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429868

  • £398.56 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(3-Bromophenyl)-2-thiohydantoin
271.13
MFCD27665230
3-(3-Bromophenyl)-2-thioxo-4-imidazolidinone

Specifications

3-(3-Bromophenyl)-2-thiohydantoin
Solid
271.13
3-(3-Bromophenyl)-2-thioxo-4-imidazolidinone
5g
95%
MFCD27665230
Videos
SDS