missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KECK™ Tubing Part for Adapter KA, with flat EPDM sealing 16 mm

Spare part for KECK™ Adapter KA.

Brand:  DWK Life Sciences 286339003

Code : A0

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 10480191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

The hose part as spare part for KECK™ Adapter KA is available in 2 sizes: Hose 4.5 and 9 mm.

  • KECK™ Adapter KA is available in 2 sizes: Hose 4.5 and 9 mm.
Specifications

Specifications

No
The hose part is a spare part for KECK™ Adapter KA
9 mm Connectors
Polypropylene, EPDM
100/Pk.
Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers