missing translation for 'onlineSavingsMsg'
Learn More

Duran™ 3.3 Borosilicate Glass Tubing Part

Manufactured from Borosilicate Glass. DURAN 3.3 Borosilicate Glass Tubing Parts are used with KA adapters.

Brand:  Duran™ 286339106

Code : A0

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 10663592

  • £3.26 / Each


Estimated Shipment
04-07-2018
Add to basket

Description

Description

  • KECK™ Tubing part for adapters KA 
  • With EPDM sealing 16 mm
Specifications

Specifications

Light Blue
Tubing Part
4mm hose connection
Duran 3.3 Borosilicate Glass
1/Pk.
Videos