missing translation for 'onlineSavingsMsg'
Learn More

(3,3,3-Trifluoropropyl)chlorodimethylsilane, 95%, Thermo Scientific Chemicals

Catalog Number 10795832
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1481-41-0
C5H10ClF3Si
190.67
KBAZUXSLKGQRJF-UHFFFAOYSA-N
trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
73866
chloro-dimethyl-(3,3,3-trifluoropropyl)silane
C[Si](C)(CCC(F)(F)F)Cl
This item is not returnable. View return policy
1481-41-0
C5H10ClF3Si
190.67
KBAZUXSLKGQRJF-UHFFFAOYSA-N
trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
73866
chloro-dimethyl-(3,3,3-trifluoropropyl)silane
C[Si](C)(CCC(F)(F)F)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.1100g/mL
Boiling Point 118°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.3770 to 1.3790 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.11
Formula Weight 190.67
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (3, 3, 3-Trifluoropropyl)chlorodimethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (3,3,3-Trifluoropropyl)chlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'