missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-[2-(Trifluoromethyl)phenyl]ureido)benzeneboronic acid pinacol ester, 95%

Brand:  Alfa Aesar™ H64653.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439056

  • £85.44 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(3-[2-(Trifluoromethyl)phenyl]ureido)benzeneboronic acid pinacol ester
406.21
MFCD28100494
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-3-(2-trifluoromethylphenyl)urea

Specifications

3-(3-[2-(Trifluoromethyl)phenyl]ureido)benzeneboronic acid pinacol ester
Solid
406.21
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-3-(2-trifluoromethylphenyl)urea
1g
95%
MFCD28100494
Videos
SDS