missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-[3-(2,5-Difluorophenyl)ureido]benzeneboronic acid pinacol ester, 95%

Brand:  Alfa Aesar™ H64429.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428916

  • £79.71 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

3-[3-(2,5-Difluorophenyl)ureido]benzeneboronic acid pinacol ester
374.19
MFCD27976896
1-(2,5-Difluorophenyl)3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea

Specifications

3-[3-(2,5-Difluorophenyl)ureido]benzeneboronic acid pinacol ester
Solid
374.19
1-(2,5-Difluorophenyl)3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea
1g
95%
MFCD27976896
Videos
SDS